Orders LinkContactUsLinkComponent
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
New Product
Leverandør:   Texwipe
TechniCloth® Flat Mop Covers
Leverandør:  Texwipe
New Product
Certifikater
TechniCloth® Flat Mop Covers
TechniCloth® Flat Mop Covers
2 tilgængelige muligheder nedenfor
Product Details & Documents
Mop covers (ISO Class 3 - 8) for cleaning and decontaminating floors.
  • Made without chemical binders or additives
  • Non-woven, economical polyester/cellulose for cleaning and removing residues
  • Chemically resistant, withstand exposure to common cleaning and disinfecting solutions
Product Family Options
Produktinformation
  • L×B
    Tilgængelighed
    Price
  • TEXWTX630
    Technicloth® floor mop wipers
    Technicloth® floor mop wipers
    250×600 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 12 Items
    1 * 12 Items - 7.750,00 kr
    7.750,00 kr
    1 * 12 Items
       7.750,00 kr
    Specifikationer:
    New Product
    New Product
  • 115-0480
    Technicloth® floor mop wipers
    Technicloth® floor mop wipers
    690×380 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 Items
    1 * 4 Items - 3.450,00 kr
    3.450,00 kr
    1 * 4 Items
       3.450,00 kr
    Specifikationer:
    New Product
    New Product