Orders LinkContactUsLinkComponent
Storage plates, 384-well, MASTERBLOCK®
Storage plates, 384-well, MASTERBLOCK®
Leverandør:   Greiner Bio-One
Storage plates, 384-well, MASTERBLOCK®
Leverandør:  Greiner Bio-One
Certifikater
Storage plates, 384-well, MASTERBLOCK®
Storage plates, 384-well, MASTERBLOCK®
2 tilgængelige muligheder nedenfor
Product Details & Documents
PP, without lid. Ideal for compound libraries and sample storage.
  • Biologically inert, resistant to commonly used solvents (such as DMSO, acetonitrile or acetone) and temperature resistant (−196 °C to +121 °C)
  • Uniform external dimensions and tight tolerances make robotic handling easier
  • Can be easily sealed with adhesive films, heat seals or CapMats
  • Alphanumeric well coding
  • Conical well shape enables precise pipetting and rounded square well design combines flexible working volume (20 to 225 µl) with reduced wicking and bubbling
Colour: Natural
Colour: Natural
Product Family Options
Produktinformation
  • Farve
    Tilgængelighed
    Price
  • 736-0151
    PP, V-bund, ikke-sterile, uden låg, dybbrønd MASTERBLOCK®
    PP, V-bund, ikke-sterile, uden låg, dybbrønd MASTERBLOCK®
    ufarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 60 Items
    1 * 60 Items - 4.890,00 kr
    4.890,00 kr
    1 * 60 Items
       4.890,00 kr
  • 738-0501
    Dybbrønd, V-bund, steril
    Dybbrønd, V-bund, steril
    ufarvet
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 60 Items
    1 * 60 Items - 6.450,00 kr
    6.450,00 kr
    1 * 60 Items
       6.450,00 kr