Microplates, 1536-well
Leverandør: Greiner Bio-One
Om denne vare
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
- HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
- Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
- Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
10 tilgængelige muligheder nedenfor
Product Details & Documents
PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or white in LUMITRAC™ or FLUOTRAC™ quality.
- HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
- Solid bottom plates available with high binding (MICROLON™ 600, LUMITRAC™ 600 or FLUOTRAC™ 600) or medium binding (LUMITRAC™ 200 or FLUOTRAC™ 200) surfaces
- Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Working volume: 3 - 10 µl
Specifications for Product Family
Specifikationer
- Catalog No:
- 392-0138
- 738-0067
- 738-0070
- 738-0068
- 738-0069
- 738-0077
- 738-0078
- 738-0080
- 738-0089
- 738-0084
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Miljøcertificering:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Specifikationer
Product Family Options
1536-well microplates, HiBase
Produktinformation
- FarveAnt. brøndeSterilTilgængelighedPrice
- 392-0138Fast bund, klar, MICROLON 600, sterilSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.392-0138Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0067Fast bund, hvid, LUMITRAC 600, sterilSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0067Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0070Fast bund, sort, FLUOTRAC 600, sterilSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0070Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0068Fast bund, hvid, LUMITRAC 200Specifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0068Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0069Fast bund, sort, FLUOTRAC 200Specifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0069Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0077µClear-bund, hvid, medium bindingSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0077Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0078µClear-bund, sort, medium bindingSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0078Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0080Fast bund, klarSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0080Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning
1536-well microplates, LoBase
Produktinformation
- FarveAnt. brøndeSterilTilgængelighedPrice
- 738-0089µClear-bund, sort, medium bindingSpecifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0089Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 738-0084Fast bund, sort, FLUOTRAC 200Specifikationer:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0084Environmentally PreferableLav fremstillingspåvirkningMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning