Microplates, 96-well
Supplier: Greiner Bio-One
About this item
PP, untreated, without lid. These microplates are ideal for storage of DNA or RNA, stock cultures, and long-term storage of active agents. Versions available sterile or non sterile and with round, conical or flat bottom well shape.
- Uniform external dimensions with 9 mm well to well spacing
- Alphanumeric well coding
- High chemical and temperature resistance (−196 to +121 °C)
- Sealable with adhesive films, heat seals or CapMats
- RNase- and DNase-free
16 Options Available Below
Product Details & Documents
PP, untreated, without lid. These microplates are ideal for storage of DNA or RNA, stock cultures, and long-term storage of active agents. Versions available sterile or non sterile and with round, conical or flat bottom well shape.
- Uniform external dimensions with 9 mm well to well spacing
- Alphanumeric well coding
- High chemical and temperature resistance (−196 to +121 °C)
- Sealable with adhesive films, heat seals or CapMats
- RNase- and DNase-free
Documents
Specifications for Product Family
Specifications
- Catalog No:
- 736-0116
- 736-0117
- 738-0006
- 738-0007
- GREI650203_100
- GREI650204_100
- GREI650205_100
- GREI650206_100
- 736-0118
- 738-0011
- GREI651261_100
- GREI651203_100
- GREI651204_100
- 736-0121
- 736-0122
- 738-0026
- Model:
- 736-0116
- 736-0117
- 738-0006
- 738-0007
- 736-0118
- 738-0011
- 736-0121
- 736-0122
- 738-0026
- Total volumen:
- 355 µl
- 355 µl
- 355 µl
- 355 µl
- 355 µl
- 355 µl
- 355 µl
- 355 µl
- 340 µl
- 340 µl
- 340 µl
- 340 µl
- 340 µl
- 392 µl
- 392 µl
- 392 µl
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Brøndprofil:
- U-bund
- U-bund
- U-bund
- U-bund
- U-bund
- U-bund
- U-bund
- U-bund
- V-bund
- V-bund
- V-bund
- V-bund
- V-bund
- Fladbundet
- Fladbundet
- Fladbundet
- Driftsvolumen:
- 50 - 300 µl
- 50 - 300 µl
- 50 - 300 µl
- 50 - 300 µl
- 50 - 300 µl
- 50 - 300 µl
- 50 - 300 µl
- 50 - 300 µl
- 50 - 335 µl
- 50 - 335 µl
- 50 - 335 µl
- 50 - 335 µl
- 50 - 335 µl
- 25 - 370 µl
- 25 - 370 µl
- 25 - 370 µl
- Miljøcertificering:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
Product Information
- BeskrivelseFarveAnt. brøndeSterilAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.736-0116Model736-0116Total volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.736-0117Model736-0117Total volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.738-0006Model738-0006Total volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.738-0007Model738-0007Total volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.GREI650203_100ModelTotal volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.GREI650204_100ModelTotal volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.GREI650205_100ModelTotal volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.GREI650206_100ModelTotal volumen355 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen50 - 300 µlMiljøcertificering - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.736-0118Model736-0118Total volumen340 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilV-bundDriftsvolumen50 - 335 µlMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.738-0011Model738-0011Total volumen340 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilV-bundDriftsvolumen50 - 335 µlMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.GREI651261_100ModelTotal volumen340 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilV-bundDriftsvolumen50 - 335 µlMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.GREI651203_100ModelTotal volumen340 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilV-bundDriftsvolumen50 - 335 µlMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - Specifications:
Lav fremstillingspåvirkning More
Specifications
Cat. No.GREI651204_100ModelTotal volumen340 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilV-bundDriftsvolumen50 - 335 µlMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - Specifications:
More
Specifications
Cat. No.736-0121Model736-0121Total volumen392 µlEnvironmentally PreferableBrøndprofilFladbundetDriftsvolumen25 - 370 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.736-0122Model736-0122Total volumen392 µlEnvironmentally PreferableBrøndprofilFladbundetDriftsvolumen25 - 370 µlMiljøcertificering - Specifications:
More
Specifications
Cat. No.738-0026Model738-0026Total volumen392 µlEnvironmentally PreferableBrøndprofilFladbundetDriftsvolumen25 - 370 µlMiljøcertificering