Orders LinkContactUsLinkComponent
Microplates, 96-well
Microplates, 96-well
   Sustainable Options Available
Supplier:   Greiner Bio-One
Microplates, 96-well
Supplier:  Greiner Bio-One
   Sustainable Options Available
Certificates
Microplates, 96-well
16 Options Available Below
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifications for Product Family
Specifications
  • Catalog No:
  • 391-3610
  • 736-0223
  • 391-3605
  • 391-3311
  • 391-3615
  • 736-0119
  • 738-0168
  • 738-0208
  • 736-0228
  • 738-0016
  • 736-0229
  • 391-3629
  • 738-0020
  • 738-0021
  • 738-0022
  • 738-0023
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
  • ISO 14001 (241002); ISO 50001 (241002)
Specifications
Product Family Options
96-well microplates with solid bottom
Product Information
  • Beskrivelse
    Anbefalet arbejdsvolumen
    Ant. brønde
    Steril
    Total volumen
    Availability
    Price
  • V-bund, klar
    40 - 200 µl
    96
    -
    234 µl
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 497,00 kr
    497,00 kr
    1 * 100 Items
       497,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    391-3610
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • V-bund, klar, steril
    40 - 200 µl
    96
    +
    234 µl
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 677,00 kr
    677,00 kr
    1 * 100 Items
       677,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    736-0223
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • U-bund, klar
    40 - 280 µl
    96
    -
    323 µl
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 1.110,00 kr
    1.110,00 kr
    1 * 100 Items
       1.110,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    391-3605
    Environmentally Preferable
    Miljøcertificering
  • U-bund, klar, steril
    40 - 280 µl
    96
    +
    323 µl
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 769,00 kr
    769,00 kr
    1 * 100 Items
       769,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    391-3311
    Environmentally Preferable
    Miljøcertificering
  • F-bund, klar
    40 - 340 µl
    96
    -
    382 µl
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 1.080,00 kr
    1.080,00 kr
    1 * 100 Items
       1.080,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    391-3615
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • F-bund, klar, steril
    40 - 340 µl
    96
    +
    382 µl
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 1.650,00 kr
    1.650,00 kr
    1 * 100 Items
       1.650,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    736-0119
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • F-bottom, clear
    40 - 340 µl
    -
    382 µl
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 196,00 kr
    196,00 kr
    1 * 50 Items
       196,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    738-0168
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • V-bottom, clear, non sterile
    40 - 200 µl
    -
    234 µl
    Log in for availability
    Log in for availability
    1 * 400 Items
    1 * 400 Items - 2.450,00 kr
    2.450,00 kr
    1 * 400 Items
       2.450,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    738-0208
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
96-well microplates with solid bottom, F-bottom (chimney well) well profile
Product Information
  • Beskrivelse
    Anbefalet arbejdsvolumen
    Ant. brønde
    Steril
    Total volumen
    Availability
    Price
  • Hvid, LUMITRAC™ 200 medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 844,00 kr
    844,00 kr
    1 * 40 Items
       844,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    736-0228
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • Hvid, LUMITRAC™ 600 høj binding, steril
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 912,00 kr
    912,00 kr
    1 * 40 Items
       912,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    738-0016
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • Sort, FLUOTRAC™ 200 medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 1.140,00 kr
    1.140,00 kr
    1 * 40 Items
       1.140,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    736-0229
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • Sort, FLUOTRAC™ 600 høj binding, steril
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 914,00 kr
    914,00 kr
    1 * 40 Items
       914,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    391-3629
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
96-well microplates with µClear® bottom, F-bottom (chimney well) well profile
Product Information
  • Beskrivelse
    Anbefalet arbejdsvolumen
    Ant. brønde
    Steril
    Total volumen
    Availability
    Price
  • Hvid, høj binding, steril
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 1.940,00 kr
    1.940,00 kr
    1 * 40 Items
       1.940,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    738-0020
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • Hvid, medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 1.950,00 kr
    1.950,00 kr
    1 * 40 Items
       1.950,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    738-0021
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • Sort, medium binding
    40 - 340 µl
    96
    -
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 1.930,00 kr
    1.930,00 kr
    1 * 40 Items
       1.930,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    738-0022
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)
      Lav fremstillingspåvirkning
  • Sort, høj binding, steril
    40 - 340 µl
    96
    +
    392 µl
    Log in for availability
    Log in for availability
    1 * 40 Items
    1 * 40 Items - 1.930,00 kr
    1.930,00 kr
    1 * 40 Items
       1.930,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    738-0023
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    ISO 14001 (241002); ISO 50001 (241002)