Microplates, 96-well
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
- Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
- Free of DNase, RNase and human DNA, and are non-pyrogenic
- Catalog No:
- 738-0032
- 738-0033
- 736-0227
- GREI655001-000
- 738-0015
- GREI655061_NOV
- 738-0017
- 391-3390
- 738-0167
- 738-0004
- 738-0005
- 738-0008
- 738-0009
- Total volumen:
- 199 µl
- 199 µl
- 382 µl
- 382 µl
- 382 µl
- 382 µl
- 392 µl
- 392 µl
- 382 µl
- 323 µl
- 323 µl
- 234 µl
- 234 µl
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Brøndprofil:
- Halvareal
- Halvareal
- F-bund (standard)
- F-bund (standard)
- F-bund (skorstensbrønd)
- F-bund (skorstensbrønd)
- U-bund
- U-bund
- V-bund
- V-bund
- Driftsvolumen:
- 15 - 175
- 15 - 175
- 25 - 340
- 25 - 340
- 25 - 340
- 25 - 340
- 40 - 280
- 40 - 280
- 40 - 200
- 40 - 200
- Miljøcertificering:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Product Family Options
- BeskrivelseArbejdsvolumenTotal volumen
- 96-brønds mikroplade, halvareal, MICROLON® 20015 - 175 µl199 µl1 * 40 Items991,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.738-0032Total volumen199 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilHalvarealDriftsvolumen15 - 175MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-brønds mikroplade, halvareal, MICROLON® 60015 - 175 µl199 µl1 * 40 Items1.390,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.738-0033Total volumen199 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilHalvarealDriftsvolumen15 - 175MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-brønds mikroplade, F-bund (standard), MICROLON® 20025 - 340 µl382 µl1 * 40 Items715,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.736-0227Total volumen382 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilF-bund (standard)Driftsvolumen25 - 340MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-well microplate, F-bottom (standard), MICROLON® 200, individually packed25 - 340 µl382 µl1 * 40 Items772,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.GREI655001-000Total volumen382 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilDriftsvolumenMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-brønds mikroplade, F-bund (standard), MICROLON® 60025 - 340 µl382 µl1 * 40 Items741,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.738-0015Total volumen382 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilF-bund (standard)Driftsvolumen25 - 340MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-well microplate, F-bottom (standard) with barcode39, MICROLON® 60025 - 340 µl382 µl1 * 40 Items460,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.GREI655061_NOVTotal volumen382 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilDriftsvolumenMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-brønds mikroplade, F-bund (skorstensbrønd), MICROLON® 20025 - 340 µl392 µl1 * 40 Items783,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.738-0017Total volumen392 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilF-bund (skorstensbrønd)Driftsvolumen25 - 340MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-brønds mikroplade, F-bund (skorstensbrønd), MICROLON® 60025 - 340 µl392 µl1 * 40 Items877,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.391-3390Total volumen392 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilF-bund (skorstensbrønd)Driftsvolumen25 - 340MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-well microplate, F-bottom, MICROLON® 600, low RAF, 2/bag25 - 340 µl382 µl1 * 50 Items1.090,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.738-0167Total volumen382 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilDriftsvolumenMiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-brønds mikroplade, U-bund, MICROLON® 20040 - 280 µl323 µl1 * 40 Items788,00 kr
Specifications
Cat. No.738-0004Total volumen323 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen40 - 280Miljøcertificering - 96-brønds mikroplade, U-bund, MICROLON® 60040 - 280 µl323 µl1 * 40 Items881,00 kr
Specifications
Cat. No.738-0005Total volumen323 µlEnvironmentally PreferableBrøndprofilU-bundDriftsvolumen40 - 280Miljøcertificering - 96-brønds mikroplade, V-bund, MICROLON® 20040 - 200 µl234 µl1 * 40 Items788,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.738-0008Total volumen234 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilV-bundDriftsvolumen40 - 200MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning - 96-brønds mikroplade, V-bund, MICROLON® 60040 - 200 µl234 µl1 * 40 Items935,00 kr
Lav fremstillingspåvirkning Specifications
Cat. No.738-0009Total volumen234 µlEnvironmentally PreferableLav fremstillingspåvirkningBrøndprofilV-bundDriftsvolumen40 - 200MiljøcertificeringISO 14001 (241002); ISO 50001 (241002)Lav fremstillingspåvirkning