Orders LinkContactUsLinkComponent
Steps, safety
Steps, safety
Sikkerhedsstige med 3 trin, 690 mm, 7,0 kg
Katalog # 139-9936
Leverandør:   WAKU GERATE
Steps, safety
Katalog # 139-9936
Leverandør:  WAKU GERATE
Steps, safety
Steps, safety
Sikkerhedsstige med 3 trin, 690 mm, 7,0 kg
Price
3.270,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Light, aluminium safety steps. Strong double rails with robust, tension and pressure resistant bracings for increased stability. Platform height corresponds to height of top step.
  • Anti-slip plastic feet
  • The large treads (36×23 cm) are covered with a non-slip rubber lining.
TÜV tested (DIN 4566 and 4567).
Certifications: TÜV tested (DIN 4566 and 4567).
Specifikationer
  • Beskrivelse:
    Sikkerhedsstige med 3 trin
  • Højde:
    690 mm
  • Vægt:
    7,0 kg