Orders LinkContactUsLinkComponent
Micro-tissue grinder set
Grinder set
  432-5063
Micro-tissue grinder set
  432-5063
Micro-tissue grinder set
Micro-tissue grinder set
Grinder set
10.270,00 kr
1 * 1 Items
A complete selection of Wheaton Micro tissue grinders conveniently packed in a lightweight protective carrying case.
Inkluderer følgende vævskværne:
- 1 ml Tenbroeck
- 0,5 ml kværn med 13 mm-425 GPI låg
- 1 ml Dounce
- 0,1 og 0,2 ml standard
- 2 ml Potter-Elvehjem
- 1 ml tilspidset
  • Beskrivelse:
    Grinder set