Heat resistant gloves
Without cuff, M, hvid/gul, 250 mm
Katalog # 112-2995
Leverandør: GANTERIE CO KG
Gå til:
Product Details & Documents
Manufactured from Nomex® fabric, using non-stop seamless process. Small loop terry towel fabric.
- Good mechanical resistance to abrasion and cuts
- Ambidextrous shape doubles glove life
- Excellent resistance to flames and small molten metal splashes
For handling sharp, hot components up to 250 °C. Ideal for welding or where there is a risk of hot projectiles.
Colour: Grey
EN 388, EN 407
Certifications: EN 388, EN 407
Specifikationer
- Handske størrelse:M
- Farve:hvid/gul
- Længde:250 mm
- Seq:01
- Model:Without cuff
- Cat. No.:112-2995
- Ambidextrous/Hånd-specifik:Ambidextrous
- EN norm:EN 388, EN 407
- Engangs/Genbrugelig:Genanvendelig