Latex handsker til renrum, BioClean Legion™
Supplier: Nitritex
Certificates
About this item
These ambidextrous cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments.
- Usterile, pudderfri, riflet overflade
- Forlænget udvaskning i rent demineraliseret vand sikrer ekstremt lave niveauer af klorider og ekstraherbare metaller
- Proteinniveau under 50 µg/g
- Hver størrelse har en unik stregkode på den udvendige pose med 50 par (EAN-13) og den udvendige æske (ITF-14), så data automatisk kan indføres i lagerstyringssystemer
Velegnet til brug i rentrum af klasse 100, ISO 5 eller M3,5.
Length: 400 mm
Colour: White
EN 455, EN 374, Cat. III; AQL 1.5
Certifications: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 5 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 5 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Packaging: 100 stk. pr. forseglet indvendige polyesterpose; en indvendig polyesterpose pr. forseglet udvendige polyesterpose; 10 udvendige polyesterposer pr. æske (1000 stk.).
Specifications
- Catalog No:
- 112-3077
- 112-3076
- 112-3075
- 112-3074
- Ambidextrous/Hånd-specifik:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Engangs/Genbrugelig:
- Engangs
- Engangs
- Engangs
- Engangs
- Med pudder/pudderfri:
- Pudderfri
- Pudderfri
- Pudderfri
- Pudderfri
- Renrum klasse:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Manchet:
- Rullet kant
- Rullet kant
- Rullet kant
- Rullet kant
- Manchet tykkelse:
- 0,11 mm
- 0,11 mm
- 0,11 mm
- 0,11 mm
- Finger tykkelse:
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- Håndflade tykkelse:
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- EN norm:
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374