Filtration funnels, Quickfit®
Leverandør: SciLabware
Gå til:
Om denne vare
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc.
Cone connection.
2 tilgængelige muligheder nedenfor
Product Details & Documents
PYREX®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc.
Cone connection.