Orders LinkContactUsLinkComponent
Wathmaker forceps, narrow tips
Wathmaker forceps, narrow tips
Fine, 110 mm
Catalog # 232-0129
Supplier:   COMACO HANDEL
Wathmaker forceps, narrow tips
Catalog # 232-0129
Supplier:  COMACO HANDEL
Wathmaker forceps, narrow tips
Wathmaker forceps, narrow tips
Fine, 110 mm
Price
145,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel 18/8. Watchmaker forceps with extra fine tips.
Specifications
  • Model:
    Fine
  • Længde:
    110 mm