Orders LinkContactUsLinkComponent
Bent forceps
Bent forceps
Bent forceps
Bent forceps
Stainless steel 18/8.
  • Pointed, curved
  • With serrated grip and tip
Product Family Options
  • Model
    Længde
  • Curved, pointed
    105 mm
  • Curved, pointed
    115 mm
    1 * 1 Items
    1 * 1 Items - 98,00 kr
    98,00 kr
    1 * 1 Items
       98,00 kr
  • Curved, pointed
    130 mm
  • Curved, pointed
    145 mm