Micro preparation forceps
Very delicate, curved tips, 105 mm
Katalog # 232-1917
Leverandør: HAMMACHER KARL
Gå til:
Product Details & Documents
Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).
- Resistant to corrosion
- Anti-magnetic
- Very delicate, curved
Specifikationer
- Model:Very delicate, curved tips
- Længde:105 mm