Orders LinkContactUsLinkComponent
Forceps for cover glasses
Forceps for cover glasses
Supplier:   BOCHEM
Forceps for cover glasses
Supplier:  BOCHEM
Certificates
Forceps for cover glasses
Forceps for cover glasses
3 Options Available Below
Product Details & Documents
Stainless steel or stainless steel with PTFE coating.
  • For easy, secure handling of cover glasses
  • With 6 mm wide tips
  • Can also be used for other laboratory equipment, such as filter discs
  • PTFE coated design with particularly smooth surface to prevent contamination and enable simple cleaning
Product Family Options
Product Information
  • Beskrivelse
    Længde
    Availability
    Price
  • Pincet til dækglas, bøjet, rustfrit stål
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 57,10 kr
    57,10 kr
    1 * 1 Items
       57,10 kr
  • Pincet til dækglas, bøjet, PTFE-belagt
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 294,00 kr
    294,00 kr
    1 * 1 Items
       294,00 kr
  • Pincet til dækglas, lige, rustfrit stål
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 67,50 kr
    67,50 kr
    1 * 1 Items
       67,50 kr