Låg til probeadaptere, Nalgene®
Probeadapterlåg, PP
Katalog # 734-5053
Leverandør: Thermo Fisher Scientific
Låg til probeadaptere, Nalgene®
Katalog # 734-5053
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
PP, hvid, med silikoneforsegling.
- Seal separates environment from inside of vessel and prevents contamination when using a probe
- Thermal resistance from −135 to +135 °C
- For Nalgene® 1 and 12 L fermenters
For use and insertion of 7 to 14 mm diameter probes into Nalgene® 1 and 12 L fermenters.
Specifikationer
- Beskrivelse:Probeadapterlåg, PP
- Cat. No.:734-5053
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning