First-aid kits
Om denne vare
The contents of the following first aid kits meet the requirements as specified in the HSE Approved Code of Practice. The code of practice states 'An employer should make an assessment of the first aid needs appropriate to the circumstance of each workplace'. To assist in selecting the appropriate kit, employee ratios are given as guidance provided by the British Healthcare Trade Association.
Delivery information: All kits are supplied with a first aid guidance leaflet.
7 tilgængelige muligheder nedenfor
Product Details & Documents
The contents of the following first aid kits meet the requirements as specified in the HSE Approved Code of Practice. The code of practice states 'An employer should make an assessment of the first aid needs appropriate to the circumstance of each workplace'. To assist in selecting the appropriate kit, employee ratios are given as guidance provided by the British Healthcare Trade Association.
Delivery information: All kits are supplied with a first aid guidance leaflet.
Specifications for Product Family
Specifikationer
- Catalog No:
- 118-0122
- 118-0130
- 118-0132
- 118-0134
- 118-0135
- 118-0121
- 118-0129
- Model:
- Standard (11 - 20 persons)
- Standard (21 - 50 persons)
- Industrial (1 - 20 persons)
- Industrial (1 - 50 persons)
- Indhold:
- 40 × Waterproof plasters, sterile
10 × individually wrapped wipes
4 × eye pads with bandages, non woven
6 × triangular bandages, non woven
12 × Safety pins
9 × sterile dressing, medium size
3 × sterile dressing, large size
2 pairs of disposable examination gloves
1 wall bracket - 60 × Waterproof plasters, sterile
20 × individually wrapped wipes
6 × eye pads with bandages, non woven
8 × triangular bandages, non woven
12 × Safety pins
12 × sterile dressing, medium size
4 × sterile dressing, large size
3 pairs of disposable examination gloves
1 wall bracket - 120 × Waterproof plasters, sterile
40 × individually wrapped wipes
12 × blue plastic fingerstalls (assorted)
4 × eye pads with bandages, non woven
6 × triangular bandages, non woven
12 × Safety pins
9 × sterile dressing, medium size
3 × sterile dressing, large size
2 × skin closure adhesives, sterile
10 × eyewash, sterile
2 × hygienic resuscitation kit
8 × Primapore wound dressings (80×80 mm)
4 × Primapore wound dressings (80×80 mm)
1 × emergency foil blanket
1 × dressing scissors
100 disposable examination gloves (pairs)
1 wall bracket - 120 × Waterproof plasters, sterile
40 × individually wrapped wipes
12 × blue plastic fingerstalls (assorted)
6 × eye pads with bandages, non woven
8 × triangular bandages, non woven
12 × Safety pins
12 × sterile dressing, medium size
4 × sterile dressing, large size
2 × skin closure adhesives, sterile
10 × eyewash, sterile
2 × hygienic resuscitation kit
10 × Primapore wound dressings (80×80 mm)
6 × Primapore wound dressings (80×80 mm)
1 × emergency foil blanket
1 × dressing scissors
100 disposable examination gloves (pairs)
1 wall bracket
Specifikationer
Product Family Options
Statutory first aid kits – low risk
Produktinformation
- TilgængelighedPrice
- 118-0122Mellemstort sæt, 11 - 20 personerSpecifikationer:
More
Specifications
Cat. No.118-0122ModelStandard (11 - 20 persons)Indhold40 × Waterproof plasters, sterile
10 × individually wrapped wipes
4 × eye pads with bandages, non woven
6 × triangular bandages, non woven
12 × Safety pins
9 × sterile dressing, medium size
3 × sterile dressing, large size
2 pairs of disposable examination gloves
1 wall bracket - 118-0130Stort sæt, 21 - 50 personerSpecifikationer:
More
Specifications
Cat. No.118-0130ModelStandard (21 - 50 persons)Indhold60 × Waterproof plasters, sterile
20 × individually wrapped wipes
6 × eye pads with bandages, non woven
8 × triangular bandages, non woven
12 × Safety pins
12 × sterile dressing, medium size
4 × sterile dressing, large size
3 pairs of disposable examination gloves
1 wall bracket
Refills for first aid kits – low risk
Produktinformation
- TilgængelighedPrice
- 118-0132Refill, til standardsætSpecifikationer:
More
Specifications
Cat. No.118-0132ModelIndhold - 118-0134Refill, til medium kitSpecifikationer:
More
Specifications
Cat. No.118-0134ModelIndhold - 118-0135Refill, til stort sætSpecifikationer:
More
Specifications
Cat. No.118-0135ModelIndhold
Statutory first aid kits – high risk
Produktinformation
- TilgængelighedPrice
- 118-0121Industrial first aid kit, 1 - 20 people, supplied in shatterproof ABS caseIndustrial first aid kit, 1 - 20 people, supplied in shatterproof ABS caseLog ind for tilgængelighed1 * 1 Each2.170,00 krSpecifikationer:
More
Specifications
Cat. No.118-0121ModelIndustrial (1 - 20 persons)Indhold120 × Waterproof plasters, sterile
40 × individually wrapped wipes
12 × blue plastic fingerstalls (assorted)
4 × eye pads with bandages, non woven
6 × triangular bandages, non woven
12 × Safety pins
9 × sterile dressing, medium size
3 × sterile dressing, large size
2 × skin closure adhesives, sterile
10 × eyewash, sterile
2 × hygienic resuscitation kit
8 × Primapore wound dressings (80×80 mm)
4 × Primapore wound dressings (80×80 mm)
1 × emergency foil blanket
1 × dressing scissors
100 disposable examination gloves (pairs)
1 wall bracket - 118-0129Industrial first aid kit, 1 - 50 people, supplied in shatterproof ABS caseIndustrial first aid kit, 1 - 50 people, supplied in shatterproof ABS caseLog ind for tilgængelighed1 * 1 Each4.810,00 krSpecifikationer:
More
Specifications
Cat. No.118-0129ModelIndustrial (1 - 50 persons)Indhold120 × Waterproof plasters, sterile
40 × individually wrapped wipes
12 × blue plastic fingerstalls (assorted)
6 × eye pads with bandages, non woven
8 × triangular bandages, non woven
12 × Safety pins
12 × sterile dressing, medium size
4 × sterile dressing, large size
2 × skin closure adhesives, sterile
10 × eyewash, sterile
2 × hygienic resuscitation kit
10 × Primapore wound dressings (80×80 mm)
6 × Primapore wound dressings (80×80 mm)
1 × emergency foil blanket
1 × dressing scissors
100 disposable examination gloves (pairs)
1 wall bracket