Stopcocks for dessicator
Catalog # 467-0078
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Beskrivelse:Needle-valve stopcock, PTFE, cone size 24/29, aperture 0-4 mm
- Pakn:1
Specifications
About this item
Standard ground NS 24/29 Stopcocks, interchangeable, for tubulated cover of desiccators.