Orders LinkContactUsLinkComponent
Chiral columns, ChiraDex®
Chiral columns, ChiraDex®
Supplier:   Merck
Chiral columns, ChiraDex®
Supplier:  Merck
Certifikater
Chiral columns, ChiraDex®
3 tilgængelige muligheder nedenfor
Product Details & Documents
ChiraDex® is a versatile HPLC column characterised by broad enantioselectivity and can be used for the separation of enantiomers of numerous different classes of substances. ChiraDex® is based on beta-cyclodextrin covalently linked to spherical particles of silica, and is ideal for chiral separation of hydrocarbons, steroids, phenol esters and derivatives, aromatic amines, heterocycles with five- to seven-membered ring. Simply composed RP-eluents can be used in most separations.
ChiraDex® LiChroCART® kolonner. LiChroCART®-kolonner kræver brug af manuCART® kolonneholder 1,51486.0001.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 1.50117.0001
  • 1.51333.0001
  • 1.51000.0001
  • pH-område:
  • 3-7,5
  • 3-7,5
  • 3-7,5
  • Surface area:
  • 300-360 m²/g
  • 300-360 m²/g
  • 300-360 m²/g
Specifikationer
Product Family Options
Produktinformation
  • Beskrivelse
    Kolonne form
    Kolonnemateriale
    Porestørrelse
    Partikelstørrelse
    Ø indv.
    Længde
    Pakkemateriale
    Availability
    Price
  • ChiraDex®
    Forkolonne
    Rustfrit stål
    100 Å
    5 µm
    4,00 mm
    4 mm
    Spherical silica
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 3.880,00 kr
    3.880,00 kr
    1 * 10 Items
       3.880,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.50117.0001
    pH-område
    3-7,5
    Surface area
    300-360 m²/g
  • ChiraDex®
    Analytisk
    Rustfrit stål
    100 Å
    5 µm
    4,00 mm
    250 mm
    Spherical silica
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 12.300,00 kr
    12.300,00 kr
    1 * 1 Items
       12.300,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.51333.0001
    pH-område
    3-7,5
    Surface area
    300-360 m²/g
  • ChiraDex® High Resolution
    Analytisk
    Rustfrit stål
    100 Å
    5 µm
    4,00 mm
    250 mm
    Spherical silica
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 12.300,00 kr
    12.300,00 kr
    1 * 1 Each
       12.300,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1.51000.0001
    pH-område
    3-7,5
    Surface area
    300-360 m²/g