Sleeve protectors, PP
hvid, 430 mm
113-0691
Non woven sleeve protectors made of PP.
- Flat elastic for tightening at each end 3 mm
- Approved for environments involving contact with food
Weight: 40 g/m²
: Compliant with CE standard governing class 1 minor hazards.
- Farve:hvid
- Længde:430 mm
- Cat. No.:113-0691
- Outfit applikationer:Food