Visitor Coats
hvid, One size
113-0667
Disposable visitor coats made from LDPE.
- With collar
- Waterproof
- Vinyl press stud fastening
Weight: 20 g
Colour: white
*One size fits all
- Tøj størrelse:One size
- Farve:hvid
- Seq:01
- Køn:Unisex
- Cat. No.:113-0667
- Vægt:20 g
- Outfit applikationer:General Purpose|Laboratory
- Lukkestørrelse:Med trykknapper