Laboratory aprons, PE
blå, 750×1250 mm
Katalog # GMEQOJC025DFS
Leverandør: GM EQUIPEMENT
Gå til:
Product Details & Documents
These disposable aprons are made of waterproof PE.
- Dirt repellent
- Waterproof
- Ideal for laboratory applications
Thickness: 25/35/50/70 µm
Colour: white or blue
Specifikationer
- Farve:blå
- Seq:06
- B×L:750×1250 mm
- Cat. No.:GMEQOJC025DFS
- Outfit applikationer:Laboratory