Bottles, wide neck, HDPE, rectangular, with thread
Leverandør: MELLERUD PLAST EMBALLATOR
Bottles, wide neck, HDPE, rectangular, with thread
Leverandør: MELLERUD PLAST EMBALLATOR
Certifikater
Gå til:
Om denne vare
HDPE, natural, without screw cap.
- Good chemical resistance
- with screw caps (1000 ml model without screw cap)
Ideal for sample collection and storage of liquids and dry substances.
7 tilgængelige muligheder nedenfor
Product Details & Documents
HDPE, natural, without screw cap.
- Good chemical resistance
- with screw caps (1000 ml model without screw cap)
Ideal for sample collection and storage of liquids and dry substances.
Product Family Options
Produktinformation
Screw caps
Produktinformation
- Katalog #BeskrivelseTilgængelighedPrice