Burette clamps, double
Klemme til 2 buretter, 2 buretter
Catalog # 241-3169
Supplier: BOCHEM
Jump to:
Product Details & Documents
Aluminium.
- With bosshead for fixing to retort stand with Ø 0 to 13
- With PVC rolls
- Distance between rolls 170 mm
Specifications
- Til:2 buretter
- Beskrivelse:Klemme til 2 buretter