Orders LinkContactUsLinkComponent
Burette Covers
Burette Covers
Burette covers, 20Ø×30H mm
Katalog # 612-9951
Leverandør:   Brand
Burette Covers
Katalog # 612-9951
Leverandør:  Brand
Burette Covers
Burette Covers
Burette covers, 20Ø×30H mm
Price
219,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Made of PP. Ribbed inside to prevent suction attachment during titration.
  • Ribbed inside to prevent suction attachment during titration
ØxH: 20x30 mm
Specifikationer
  • Beskrivelse:
    Burette covers, 20Ø×30H mm