Orders LinkContactUsLinkComponent
Automatic burettes, Sofnol
Automatic burettes, Sofnol
Supplier:   SciLabware
Automatic burettes, Sofnol
Supplier:  SciLabware
Certificates
Automatic burettes, Sofnol
Automatic burettes, Sofnol
3 Options Available Below
Product Details & Documents
Borosilicate glass.
  • Rotaflo® PTFE stopcock for grease-free performance and fine drop-wise control
  • Graduations in white enamel
  • Automatic zeroing and easy filling
Ordering information: Burettes and bottles must be ordered separately.
Product Family Options
Product Information
  • Kapacitet
    Inddeling
    Tolerance
    Availability
    Price
  • 10 ml
    0,1 ml
    0,01 ± ml
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 5.060,00 kr
    5.060,00 kr
    1 * 5 Items
       5.060,00 kr
  • 20 ml
    0,1 ml
    0,01 ± ml
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 5.380,00 kr
    5.380,00 kr
    1 * 5 Items
       5.380,00 kr
  • 30 ml
    0,1 ml
    0,01 ± ml
    Log in for availability
    Log in for availability
    1 * 5 Items
    1 * 5 Items - 5.710,00 kr
    5.710,00 kr
    1 * 5 Items
       5.710,00 kr
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 612-2909
    Ekstra reservoirflaske, 1000 ml, ØxH: 95x195 mm
    Log in for availability
    1 * 1 Items - 245,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 612-2908
    Ekstra reservoirflaske, 500 ml, ØxH: 75x160 mm
    Log in for availability
    1 * 1 Items - 157,00 kr