Brush for butyrometer
Naturbørster, Wire, 260 mm
Katalog # HECH91120000012
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Brush with natural bristles and two brush parts, outer 22×12 mm, inner 60×45 mm. Galvanized iron thread handle.
Specifikationer
- Børstemateriale:Naturbørster
- Handle material:Wire
- Samlet længde:260 mm