Test Tube Rack, Polystyrene
Storage box, -, 150×150×45 mm
Katalog # 212-0285
Leverandør: SAVEEN BIOTEC
Gå til:
Product Details & Documents
Polystyrene.
Test tube rack for 5×10 test tubes.
Hole size 12 mm. Length 146 mm, width 73 mm, height 40 mm.
Storage box for 2 test tube racks. Made from grey cardboard with overlapping lid. Size: 150×150×45 mm.
Specifikationer
- Ant. huller:-
- Beskrivelse:Storage box
- Dimensioner:150×150×45 mm