Orders LinkContactUsLinkComponent
Cryoboxes, Nalgene®
Cryoboxes, Nalgene®
   Sustainable Options Available
Cryoboxes, Nalgene®
   Sustainable Options Available
Certificates
Cryoboxes, Nalgene®
3 Options Available Below
Limited Time Sale
Select items below are currently on sale
Product Details & Documents
Hvid PC, autoklaverbar.
  • Durable, all-plastic CryoBoxes™ for short- or long-term storage
  • Plastic boxes are autoclavable for decontamination
  • Printed grid on plastic lids for easy inventory management
  • Chipboard/cardboard boxes have dividers and vents in base for good drainage and air circulation
  • Available with dividers from 25 to 169 tube storage
Til opbevaring af cryorør ved ultralave temperaturer (−196 til +121 °C).
Transparent låg med påtrykt gitter giver overblik over indholdet og er udformet til at undgå forkert påsætning.
Der kan bruges cryo-penne til at skrive på låget.
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes. Choose from a variety of durable plastic Thermo Scientific™ CryoBox™ styles with keyed lids and printed location grids, or select economical cardboard/chipboard CryoBoxes™ for short-term storage. The plastic boxes are optimised to fit our own brand of Nalgene™ and Nunc™ vials and microcentrifuge tubes best.
Specifications for Product Family
Specifications
  • Catalog No:
  • 479-3311
  • 479-5131
  • 479-0011
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
  • Beskrivelse
    Rum
    Til
    Farve
    B×D×H udv.
    Availability
    Price
  • Nalgene® polycarbonate cryobox, weight: 0,52 kg
    5×5
    Cryogenic vials 1,2 and 2,0 ml
    hvid
    76×76×52 mm
    Log in for availability
    Log in for availability
    1 * 8 Items
    1 * 8 Items - kr462.00
    kr462.00
    Sale
    Valid until 31/03/2026
    SaleValid until 31/03/2026
    1 * 8 Items
       kr462.00
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    479-3311
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Lav fremstillingspåvirkning
  • Nalgene® polycarbonate cryobox, weight : 4 kg
    9×9
    Cryogenic vials 1,2 and 2,0 ml
    hvid
    133×133×52 mm
    Log in for availability
    Log in for availability
    1 * 4 Items
    1 * 4 Items - kr365.00
    kr365.00
    Sale
    Valid until 31/03/2026
    SaleValid until 31/03/2026
    1 * 4 Items
       kr365.00
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    479-5131
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Lav fremstillingspåvirkning
  • Nalgene® polycarbonate cryobox, weight : 0,83 kg
    9×9
    Cryogenic vials 5,0 ml
    hvid
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 4 Items
    1 * 4 Items - kr458.00
    kr458.00
    Sale
    Valid until 31/03/2026
    SaleValid until 31/03/2026
    1 * 4 Items
       kr458.00
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    479-0011
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Lav fremstillingspåvirkning