Orders LinkContactUsLinkComponent
Cool Boxes
Cool Boxes
Cool Boxes
Cool Boxes
Blue-white PP. Sturdy, robust design, ideal for storing and transporting temperature-sensitive materials and samples.
  • Box casing and lid fully insulated with CFC-free PU foam
  • A rubber seal ensures the boxes are airtight, a lid with non breakable handle can be securely locked
  • Excellent cooling performance (depending on the model up to 24 hours cooling)
Product Family Options
  • Beskrivelse
    Kapacitet
    Dimensioner
  • Cooling box ISOTHERM Extreme
    10 l
    310×225×350 mm (ext); 265×165×270 mm (int)
    1 * 1 Items
    1 * 1 Items - 414,00 kr
    414,00 kr
    1 * 1 Items
       414,00 kr
  • Cooling box ISOTHERM Extreme
    17 l
    365×295×450 mm (ext); 320×195×310 mm (int)
    1 * 1 Items
    1 * 1 Items - 479,00 kr
    479,00 kr
    1 * 1 Items
       479,00 kr