Flasker, bred hals, rektangulære, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Certifikater
Om denne vare
HDPE, gennemskinnelig med PP skruelåg eller PPCO, gennemskinnelig med PP skruelåg.
- HDPE: Compatibility with most alcohols, bases and acids
- PPCO: Very good chemical resistance, heat resistant up to 121 °C, autoclavable
- Extra wide opening
Large volume multipurpose bottles for a wide range of applications. Space saving, with convenient integral handles.
Specifikationer
- Catalog No:
- 215-7382
- 215-7372
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Miljøcertificering:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifikationer
Relateret information
Frequently Bought Together