Orders LinkContactUsLinkComponent
Bottle, narrow neck, with thread
Bottle, narrow neck, with thread
Supplier:   REXAM
Bottle, narrow neck, with thread
Supplier:  REXAM
Certificates
Bottle, narrow neck, with thread
Bottle, narrow neck, with thread
3 Options Available Below
Product Details & Documents
Soda-lime glass, brown, narrow without screw caps.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
DIN thread. Brown coloured to protect light-sensitive materials and supplied with dust protection cap. Space-saving (square shape) for storage and transport. Stable, non-slip.
Closures in various designs are available on request (see Cat. No. 217-9200 to 217-9203 for screw caps and 217-0202 to 217-0205 for pouring rings)).
Product Family Options
Product Information
  • Kapacitet
    Gevind
    Udv.Ø
    Højde
    B×D×H
    Availability
    Price
  • 250 ml
    GL 32
    63×63×155 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 22.860,00 kr
    22.860,00 kr
    1 * 1 Items
       22.860,00 kr
  • 500 ml
    GL 32
    77×77×186 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 30,60 kr
    30,60 kr
    1 * 1 Items
       30,60 kr
  • 2500 ml
    GL 45
    139 mm
    283 mm
    Log in for availability
    Log in for availability
    1 * 11 Items
    1 * 11 Items - 745,00 kr
    745,00 kr
    1 * 11 Items
       745,00 kr