Orders LinkContactUsLinkComponent
Dropping bottles, glass
Dropping bottles, glass
Dropping bottles, glass
Dropping bottles, glass
Glass, amber, with screw cap, pipette and rubber suction bulb.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
Brown coloured to protect light-sensitive materials.
Product Family Options
  • Kapacitet
    Højde
    Udv.Ø
  • 10 ml
    62 mm
    25 mm
    1 * 1 Items
    1 * 1 Items - 24,40 kr
    24,40 kr
    1 * 1 Items
       24,40 kr
  • 25 ml
    80 mm
    33 mm
  • 50 ml
    92 mm
    38 mm
  • 100 ml
    110 mm
    47 mm
Accessories
  •  
    Beskrivelse
    Til
  • 215-1876
    Screw cap, pipette and rubber suction bulb
    10 ml dropping bottles
    1 * 1 Items - 33,50 kr