Orders LinkContactUsLinkComponent
Bottle/carboys, rectangular, with handles, HDPE
Bottle/carboys, rectangular, with handles, HDPE
Bottle/carboys, rectangular, with handles, HDPE
Certificates
Bottle/carboys, rectangular, with handles, HDPE
Bottle/carboys, rectangular, with handles, HDPE
3 Options Available Below
Product Details & Documents
HDPE, with or without screw cap. Rectangular, with handles. Threaded, 50 and 100 mm.
Ordering information: The 5 and 10 liter jugs ​​with 100mm opening are intended for use with dry products. If the jug is to be used with liquid, a foil or similar material must be used to open the bottle. Applies to product numbers MELP48C0500100117D and 215-3064.
Product Family Options
Product Information
  • Beskrivelse
    Kapacitet
    Hals-Ø
    Availability
    Price
  • Jumper jar with white lid narrow neck
    2,5 l
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr64.10
    kr64.10
    1 * 1 Items
       kr64.10
  • Jumper jar with white lid narrow neck
    5 l
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr97.10
    kr97.10
    1 * 1 Items
       kr97.10
  • Plastic bottle, cap
    5 l
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - kr126.00
    kr126.00
    1 * 1 Items
       kr126.00
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 215-3052
    Lock, svart, PE, med gänga 50 mm
    Log in for availability
    1 * 1 Items - 10,30 kr
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 215-3053
    Spigot for plastic carboy, 2 to 10 L
    Log in for availability
    1 * 1 Items - 90,00 kr