Double bosshead
Leverandør: R L ENTERPRISES
Gå til:
Om denne vare
Bossheads made from nickel plated cast alloy.
- Either with or without blue plastic headed nickel plated screws
- For use with rods up to 16 mm
2 tilgængelige muligheder nedenfor
Product Details & Documents
Bossheads made from nickel plated cast alloy.
- Either with or without blue plastic headed nickel plated screws
- For use with rods up to 16 mm
Product Family Options
Produktinformation
- MaterialeTil stang ØTilgængelighedPrice
- 241-7867Bosshead with blue plastic headed screwBosshead with blue plastic headed screwStøbt legering16 mmLog ind for tilgængelighed1 * 1 Items244,00 kr
- RLAA2005/1PLBosshead without blue plastic headed screwBosshead without blue plastic headed screwStøbt legering16 mmLog ind for tilgængelighed1 * 1 Items197,00 kr