Orders LinkContactUsLinkComponent
Double bosshead
Double bosshead
Thumb screw, Aluminiumslegering, 16,5 mm
Katalog # 241-3168
Leverandør:   BOCHEM
Double bosshead
Katalog # 241-3168
Leverandør:  BOCHEM
Double bosshead
Double bosshead
Thumb screw, Aluminiumslegering, 16,5 mm
Price
103,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Double bosshead made from aluminium.
Screw type: Thumb screw, M8 (Type 1).
Certifications: DIN 12895
Specifikationer
  • Beskrivelse:
    Thumb screw
  • Materiale:
    Aluminiumslegering
  • Til stang Ø:
    16,5 mm