Combustion boats
80×13×9 mm
HALDLSCH/80X13X9/U
Boats, made of porcelain, unglazed, with eyelet to aid recovery from furnace.
- Temperature resistance up to 1350 °C
- Very good chemical resistance
- B×D×H:80×13×9 mm
Boats, made of porcelain, unglazed, with eyelet to aid recovery from furnace.