Waste Bin
grå, 60 L, 360×260×610 mm
Katalog # 129-9922
Leverandør: Zellstoff-Vertriebs-GmbH & Co. KG
Gå til:
Product Details & Documents
Coated steel wire.
This basket can be placed on the floor or mounted on the wall. For use with waste disposal bags.
Specifikationer
- Kapacitet:60 L
- Farve:grå
- B×D×H:360×260×610 mm