Orders LinkContactUsLinkComponent
Beakers, low form, Nalgene®
Beakers, low form, Nalgene®
   Bæredygtige muligheder
Leverandør:   Thermo Fisher Scientific
Beakers, low form, Nalgene®
   Bæredygtige muligheder
Certifikater
Beakers, low form, Nalgene®
10 tilgængelige muligheder nedenfor
Product Details & Documents
PPCO, translucent.
  • Very good chemical resistance
  • Heat resistant up to +121 °C, autoclavable at +121 °C (max. 20 minutes)
  • Smooth surfaces for easy, fast cleaning
Designed for general laboratory use. With spout and easy to read printed graduations with size code, plastic code, maximum temperature for use and 'Not flameproof' symbol. Ribbed for easy stacking. No drip, single spout design provides precision pouring control.
ISO 7056
Certifications: ISO 7056
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 213-7100
  • 213-7101
  • 213-7102
  • 213-7103
  • 213-7104
  • 213-7105
  • 213-7106
  • 213-7107
  • 213-7108
  • 213-7109
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • Miljøcertificering:
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
  • ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
Specifikationer
Product Family Options
Produktinformation
  • Kapacitet
    Graduation rækkevidde
    Tilgængelighed
    Price
  • 30 ml
    5 - 30 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 12 Items
    1 * 12 Items - 382,00 kr
    382,00 kr
    1 * 12 Items
       382,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7100
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 50 ml
    10 - 50 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 12 Items
    1 * 12 Items - 412,00 kr
    412,00 kr
    1 * 12 Items
       412,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7101
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 100 ml
    10 - 100 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 12 Items
    1 * 12 Items - 452,00 kr
    452,00 kr
    1 * 12 Items
       452,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7102
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 150 ml
    20 - 50 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 12 Items
    1 * 12 Items - 517,00 kr
    517,00 kr
    1 * 12 Items
       517,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7103
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 250 ml
    25 - 250 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 6 Items
    1 * 6 Items - 361,00 kr
    361,00 kr
    1 * 6 Items
       361,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7104
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 400 ml
    50 - 400 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 6 Items
    1 * 6 Items - 466,00 kr
    466,00 kr
    1 * 6 Items
       466,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7105
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 600 ml
    100 - 600 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 4 Items
    1 * 4 Items - 401,00 kr
    401,00 kr
    1 * 4 Items
       401,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7106
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 1000 ml
    100 - 1000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 3 Items
    1 * 3 Items - 382,00 kr
    382,00 kr
    1 * 3 Items
       382,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7107
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 2000 ml
    200 - 2000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 274,00 kr
    274,00 kr
    1 * 1 Items
       274,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7108
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
  • 4000 ml
    500 - 4000 ml
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 476,00 kr
    476,00 kr
    1 * 1 Items
       476,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    More
    Specifications
    Cat. No.
    213-7109
    Environmentally Preferable
    Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse
    Miljøcertificering
    ISO14001 (Certificate Registration No. EMS 752603), ACT Label
    NAR Certificate of Retirement
      Lav fremstillingspåvirkning|Produktgennemsigtighed og videregivelse