Orders LinkContactUsLinkComponent
Wire basket
Wire basket
400×300×200 mm
Katalog # 216-7470
Leverandør:   HUPFER METALLWERKE
Wire basket
Katalog # 216-7470
Leverandør:  HUPFER METALLWERKE
Wire basket
Wire basket
400×300×200 mm
Price
2.600,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel, electrolytically polished.
  • Very good chemical and thermal resistance
  • Foldaway carrying handle with plastic handgrip
  • Robust design (wire mesh size 8×8 mm) with two rods for base reinforcement
This rectangular baskets are suitable for a variety of uses.
Specifikationer
  • B×D×H:
    400×300×200 mm