Orders LinkContactUsLinkComponent
Multiple adapters
Multiple adapters
Supplier:   Lenz Laborglas
Multiple adapters
Supplier:  Lenz Laborglas
Certificates
Multiple adapters
Multiple adapters
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3.
  • Meget god kemikalieresistens
  • Høj temperatur modstand
  • Minimal termisk ekspansion giver relativt høj modstand mod temperaturændringer
Med to standard jord 29/32 fatninger og en standard jord 29/32 kegle. Sidehalsen skrå opad i en vinkel på 105°.
Documents
Product Family Options
Product Information
  • Konusstr.
    Kappestr.
    Availability
    Price
  • 14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 251,00 kr
    251,00 kr
    1 * 1 Items
       251,00 kr
  • 19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 273,00 kr
    273,00 kr
    1 * 1 Items
       273,00 kr
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 296,00 kr
    296,00 kr
    1 * 1 Items
       296,00 kr
  • 29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 358,00 kr
    358,00 kr
    1 * 1 Items
       358,00 kr