Chambered cover glasses, Nunc™, Lab-Tek™
Katalog # 732-3644
Leverandør: Thermo Fisher Scientific
Specifikationer
- Beskrivelse:Coverglass for 16-well chamber slide
- L×B:22×47 mm
- Environmentally Preferable:Lav fremstillingspåvirkning
- Miljøcertificering:NAR Certificate of Retirement
- Pakn:96
Specifikationer
Om denne vare
These cover glasses provide secure content on a slide while viewing under a microscope.
For use with slide flask (NUNC170920) and Thermo Scientific™ Flaskette™ products.