Vaskeflasker, bred åbning, farvekodede, Unitary™, Nalgene®
500 ml, 38-415
Katalog # NALG2423-0500
Leverandør: Thermo Fisher Scientific
Vaskeflasker, bred åbning, farvekodede, Unitary™, Nalgene®
Katalog # NALG2423-0500
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
LDPE, translucent, with PP closure.
- Good chemical resistance
- Good temperature resistance
Coded Unitary LDPE wash bottles, which come in a convenient, pre-packaged and easy-to-identify assortment. Red, white, blue or yellow closures/stems on these flexible unitary bottles permit easy content identification.
Specifikationer
- Gevind:38-415
- Kapacitet:500 ml
- Miljøcertificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lav fremstillingspåvirkning
- Cat. No.:NALG2423-0500