Ventilationsfilter til dunke, Nalgene®
0,2 µm, 300 cm²
Katalog # 517-0001
Leverandør: Thermo Fisher Scientific
Ventilationsfilter til dunke, Nalgene®
Katalog # 517-0001
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
These vent filters with hydrophobic PTFE membrane and PP housing are designed to maintain purified water stored in carboys. They can also be used for venting and solvent filtration applications.
- Use up to five times in venting applications
- 6 ml liquid hold-up volume
Specifikationer
- Filtreringsområde:300 cm²
- Porestørrelse:0,2 µm
- Seq:01
- Cat. No.:517-0001
- Miljøcertificering:NAR Certificate of Retirement
- Environmentally Preferable:Lav fremstillingspåvirkning