Orders LinkContactUsLinkComponent
Filter Holder Accessories
Filter Holder Accessories
Supplier:   Merck
Filter Holder Accessories
Supplier:  Merck
Certificates
Filter Holder Accessories
Filter Holder Accessories
29 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Beskrivelse
    Til
    Availability
    Price
  • Patch test kit, contains PVC bottle (for sample collection), PVC bottle (for solvent dispensing), aluminum assembly (for filter holder), polyethylene bottle (for sample collection), polyethylene bottle (for solvent dispensing), stainless steel assembly (for filter holder)
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 23.530,00 kr
    23.530,00 kr
    1 * 1 Each
       23.530,00 kr
  • Hex nipple, 6,35 mm NPTM 51 mm, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 973,00 kr
    973,00 kr
    1 * 1 Items
       973,00 kr
  • ¹/₄" HB to M Luer lock nickel/chrome plated
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 278,00 kr
    278,00 kr
    1 * 1 Items
       278,00 kr
  • Pressure tubing with 6,35 mm NPT adapters, PE
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.690,00 kr
    1.690,00 kr
    1 * 1 Items
       1.690,00 kr
  • Tubing, 6 mm, O.Ø 3 m, polypropylene
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 661,00 kr
    661,00 kr
    1 * 1 Items
       661,00 kr
  • O-ring viton
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 692,00 kr
    692,00 kr
    1 * 6 Items
       692,00 kr
  • Swinnex 25 mm with back-pressure screen, polypropylene housing, female Luer-Lok® inlet connection male Luer outlet slip
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 393,00 kr
    393,00 kr
    1 * 1 Items
       393,00 kr
  • Tube adapter, 6,35 mm NPTM to 6,35 mm tube, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 661,00 kr
    661,00 kr
    1 * 1 Items
       661,00 kr
  • Handle and valve assembly
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.920,00 kr
    5.920,00 kr
    1 * 1 Items
       5.920,00 kr
  • Clamp (hose) 12,7 mm (0,43 - 0,810), stainless steel
    Log in for availability
    Log in for availability
    1 * 4 Items
    1 * 4 Items - 325,00 kr
    325,00 kr
    1 * 4 Items
       325,00 kr
  • Street elbow, 6,35 mm NPTF to M, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 785,00 kr
    785,00 kr
    1 * 1 Items
       785,00 kr
  • Pipe hread fitting 6,35 mm NPTF tee, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 755,00 kr
    755,00 kr
    1 * 1 Items
       755,00 kr
  • Tubing 20 psi silicon 3048 mm, Int.Ø 12,7 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.500,00 kr
    2.500,00 kr
    1 * 1 Items
       2.500,00 kr
  • Hose connector 6,35 mm NPTF quick-release nipple,
    stainless steel
    PL-1 Cartridge Housings
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 592,00 kr
    592,00 kr
    1 * 1 Items
       592,00 kr
  • Tubing for vacuum, 3,175 mm, fluoroelastomer, 190 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 405,00 kr
    405,00 kr
    1 * 1 Items
       405,00 kr
  • PVC bottle, 125 ml
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 209,00 kr
    209,00 kr
    1 * 1 Items
       209,00 kr
  • Stainless steel holder support with base, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.810,00 kr
    8.810,00 kr
    1 * 1 Items
       8.810,00 kr
  • Solvent dispensing bottle, polyethylene, 66 mm height
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 357,00 kr
    357,00 kr
    1 * 1 Items
       357,00 kr
  • Tygon tubing, int.Ø 4.76 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 241,00 kr
    241,00 kr
    1 * 1 Items
       241,00 kr
  • Hose connector, 3,175 mm NPTF quick-release valve
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.130,00 kr
    3.130,00 kr
    1 * 1 Items
       3.130,00 kr
  • Pipe thread valve 3,175 - 3,175 mm, NPTM 3-way plug valve
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.440,00 kr
    7.440,00 kr
    1 * 1 Items
       7.440,00 kr
  • Replacement part kit for hand vacuum pump, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.050,00 kr
    1.050,00 kr
    1 * 1 Items
       1.050,00 kr
  • Hand vacuum pump, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.740,00 kr
    3.740,00 kr
    1 * 1 Items
       3.740,00 kr
  • Adapter tubing to male Luer
    Log in for availability
    Log in for availability
    1 * 10 Items
    1 * 10 Items - 242,00 kr
    242,00 kr
    1 * 10 Items
       242,00 kr
  • Vent/relief valve, 9 bar, Int.Ø 6,3 mm
    293 mm stainless steel sanitary filter holder
    PL-1 cartridge housings
    for use with PS-1 cartridge housings
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.320,00 kr
    2.320,00 kr
    1 * 1 Items
       2.320,00 kr
  • Remote sampling assembly, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.100,00 kr
    3.100,00 kr
    1 * 1 Items
       3.100,00 kr
  • Sampler vlave hose assembly, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 15.290,00 kr
    15.290,00 kr
    1 * 1 Items
       15.290,00 kr
  • Cylinder O-rings, fluoroelastomer
    ZHE hazardous waste filtration system
    Log in for availability
    Log in for availability
    1 * 3 Items
    1 * 3 Items - 1.860,00 kr
    1.860,00 kr
    1 * 3 Items
       1.860,00 kr
  • Connection adapter 6,35 mm NPTF to 6,35 mm Int.Ø hose nylon
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 204,00 kr
    204,00 kr
    1 * 1 Items
       204,00 kr