MultiScreen® Collection Plates, 96 Well
klar, 100 μl sample volume (per well), 96, -
Katalog # MILFMATRNPS50
Leverandør: Merck
Gå til:
Product Details & Documents
These 96 well collection plates are suitable for mammalian cell culture and parallel artificial membrane permeation assay (PAMPA).
- Transport receiver plate
- Clear
- Non sterile
Made of polystyrene
Specifikationer
- Ant. brønde:96
- Steril:-
- Farve:klar
- Brøndvolumen:100 μl sample volume (per well)