Orders LinkContactUsLinkComponent
Millex® Polyethersulfone Inline Filters
Millex® Polyethersulfone Inline Filters
Leverandør:   Merck
Millex® Polyethersulfone Inline Filters
Leverandør:  Merck
Certifikater
Millex® Polyethersulfone Inline Filters
2 tilgængelige muligheder nedenfor
Product Details & Documents
The bi-directionally supported Millex-GP50 filter is a sterilising filter for aqueous solutions. It removes microorganisms, particles, precipitates, and undissolved powders larger than 0,22 μm. This single-use product consists of a 0,22 μm membrane filter sealed in an acrylic housing.
  • Sterile filtration of tissue culture media, additives, buffers, and water
  • Sterile, gamma irradiated
  • Hydrophilic
  • Millex-GP50 filter is non-pyrogenic and non-toxic
For use with peristaltic pump, a pump drive and pump head that delivers a constant speed of 60 revolutions per minute (rpm) is recommended. This generates an average flow rate of 100 ml/min. Recommended tubing: 4 to 9,5 mm Int. Ø. silicone rubber or latex tubing.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • MILFSLGP05010
  • MILFSLGPB5010
  • Housing material:
  • Methacrylate-butadiene-styrene (MBS)
  • Methacrylate-butadiene-styrene (MBS)
  • Indløb/udløb forbindelse:
  • 96 mm, including bell
  • 96 mm, including bell
  • Maks. tryk:
  • 3,3 bar
  • 3,3 bar
Specifikationer
Product Family Options
Produktinformation
  • Porestørrelse
    Filtreringsområde
    Ø
    Steril
    Tilgængelighed
    Price
  • MILFSLGP05010
    Inline filter, 6,35 - 12,7 mm hose barb with Female Luer slip interior
    Inline filter, 6,35 - 12,7 mm hose barb with Female Luer slip interior
    0,22 μm
    19,6 cm²
    50 mm
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 1.463,66 kr
    1.463,66 kr
    1 * 10 Items
       1.463,66 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MILFSLGP05010
    Housing material
    Methacrylate-butadiene-styrene (MBS)
    Indløb/udløb forbindelse
    96 mm, including bell
    Maks. tryk
    3,3 bar
  • MILFSLGPB5010
    Inline filter, 6,35 - 12,7 mm stepped hose barb with Female Luer slip interior, outlet fitting bell
    Inline filter, 6,35 - 12,7 mm stepped hose barb with Female Luer slip interior, outlet fitting bell
    0,22 μm
    19,6 cm²
    50 mm
    +
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 10 Items
    1 * 10 Items - 1.650,00 kr
    1.650,00 kr
    1 * 10 Items
       1.650,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    MILFSLGPB5010
    Housing material
    Methacrylate-butadiene-styrene (MBS)
    Indløb/udløb forbindelse
    96 mm, including bell
    Maks. tryk
    3,3 bar