Rotary Agitator
Rotary agitator, 115 V/60 Hz
Katalog # MILFYT310RAHW
Leverandør: Merck
Gå til:
Product Details & Documents
If your waste samples contain non-volatile or semi-volatile substances or metals, use this hazardous waste filtration system for saparation, extratction, and filtration.
- Contains interior holder with PTFE coating to prevent heavy metal contamination
- Allows easy introduction of bulky samples through wide entry port and removable top plate
- Disassembles for easy cleaning
Developed in collaboration with the US Environmental Protection Agency (EPA), the system is designed specifically for the US EPA Extraction Procedure (EP) Toxicity Test and Toxicity Characteristic Leaching Procedure (TCLP). The device separates solid and liquid phases of waste samples for analysis.
Specifikationer
- Beskrivelse:Rotary agitator, 115 V/60 Hz