Orders LinkContactUsLinkComponent
Buffered NaCl Peptone Solution pH 7,0
Buffered NaCl Peptone Solution pH 7,0
Leverandør:   LIOFILCHEM
Buffered NaCl Peptone Solution pH 7,0
Leverandør:  LIOFILCHEM
Certifikater
2 tilgængelige muligheder nedenfor
Product Details & Documents
Buffered NaCl Peptone Solution pH 7,0 is used for dissolving, suspending and diluting test samples. Its composition conforms to recommendations in the Harmonised Pharmacopoeias for the microbiological examination of non-sterile products.
Certifications: This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics.
Ordering information: For professional use only.
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • LIOF402730
  • LIOF403410
  • Opbevaringstemperatur:
  • 10...25 °C
  • 10...25 °C
Specifikationer
Product Family Options
Produktinformation
  • pH-værdi
    Påfyldning
    Pakn
    Tilgængelighed
    Price
  • LIOF402730
    Buffered NaCl peptone solution
    Buffered NaCl peptone solution
    7,0
    90 ml
    6
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 6 Items
    1 * 6 Items - 306,00 kr
    306,00 kr
    1 * 6 Items
       306,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    LIOF402730
    Opbevaringstemperatur
    10...25 °C
  • LIOF403410
    Buffered NaCl peptone solution
    Buffered NaCl peptone solution
    7,0
    300 ml
    6
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 6 Items
    1 * 6 Items - 418,00 kr
    418,00 kr
    1 * 6 Items
       418,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    LIOF403410
    Opbevaringstemperatur
    10...25 °C