Ingredient Bin Slant
20,5 gallons / 325 cups, 12,75 kg, hvid
Katalog # RUBBFG360088WHT
Leverandør: Rubbermaid Commercial Products
Gå til:
Product Details & Documents
The Rubbermaid commercial ProSave® ingredient and food storage mobile bin is a bulk food storage container on wheels. With a slanted front, sliding opening, and 32-ounce scoop, these food storage containers make it easy to transport ingredients.
- Integrated lid for easy access
- 32-ounce scoop included
- Internal scoop hanger for food safety
- Rectangular design for space saving
- Measures W×L×H: 13¹/₈×29¹/₄×28"
Specifikationer
- Kapacitet:20,5 gallons / 325 cups
- Farve:hvid
- Vægt:12,75 kg
- Cat. No.:RUBBFG360088WHT
- Materiale:PP